missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ GL 18 Pressure Equalization Set

Product Code. 11850580 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
16 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
11850580 16 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11850580 Supplier DWK Life Sciences Supplier No. 1160167
  • ONLINE EXCLUSIVE PRICE
    £74.04
    £26.51 / Pack of 1
    Save £47.53
    64% Off
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The DURAN™ GL 18 Pressure Equalization Set is an accessory for the GLS 80 four-port screw cap connection system, enabling sterile pressure equalization via membrane filter and safe closed-system liquid media transfer.

  • Pressure equalization kit for the GLS 80 four-port (GL 18 thread) screw cap connection system
  • Enables sterile pressure equalization through a membrane filter
  • Supports safe transfer of liquid media within a closed and sterile system, reducing evaporation
  • Compatible with five hose diameters: 3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm, and 12.0 mm
  • Unused ports can be fitted with a GL 18 blind cap
  • Central NS 29/32 ground joint fitting for tight closure
  • Flexible modular system constructed from PP and PTFE
  • Compatible with all GLS 80 threads for versatile use with DURAN laboratory and media bottles

Specifications

Color Red
Description Pressure equalisation cap set for 4-port, GL 18
For Use With (Application) Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor
Material Polypropylene, PTFE
Quantity 1 Ea.
Sterility Non-sterile
Thread Size (Metric) 16 mm
Neck Finish 16 mm
Diameter (Metric) 87 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Bottle Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.