Learn More
DWK Life Sciences DURAN™ Flat Flange Lid, 4 standard ground necks, with side necks: 2 x NS 29/32 angled; 1 x 14/23 parallel,, center neck: NS 29/32
Product Code. 10749101 Shop All DWK Life Sciences Products
Change view
Click to view available options
Description:
DN 100
DN 120
DN 150
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10749101 DN 100 Each
10211811 DN 150 Each
10709681 DN 120 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10749101 Supplier DWK Life Sciences Supplier No. 243924702
  • ONLINE EXCLUSIVE PRICE
    £300.30
    £294.01 / Pack of 1
    Save £6.29
    2% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The lid of a plane fan vessel is suitable for closing but also for combination with other ground joint components (e.g. dropping funnel, stirring shafts, etc.) due to the standard ground joints.
Flat flange vessels are large volume reaction vessels which are regarded as an alternative to the round flasks and consist of a bottom part and a lid. These two parts are held together by means of a clamping ring. A seal or grinding grease prevent the grinding surfaces from tilting. The flat flange vessel can be combined with other components (e.g. dropping funnels, stirring shafts, ...) by means of various standard ground joints in the lid. Flat flange vessels are well suited for preparative work such as general reactions, distillations, evaporation and drying of solid and viscous substances. Due to their walls, flat flange vessels are suitable for safe working under pressure and vacuum.
  • With one middle neck: 29/32 and three side necks (standard ground): 3 x 29/32 parallel
  • Pressure and vacuum resistant due to the wall thickness and geometry
  • Tight closure due to standard ground joint
Specifications
Description DN 100
Material Borosilicate Glass (3.3)
For Use With (Application) Chemically and thermally demanding processes; Safe working under pressure or vacuum
Neck Style 2 x NS 29/32 angled; 1 x NS 14/23 parallel
No. per Pack 1/Pk.
Socket Joint NS 29/32
Autoclavable Autoclavable