Learn More
DWK Life Sciences DURAN™ Flat Bottom Flask, narrow neck, with beaded rim
Flat base means flasks can be set down without a supporting ring.
£63.75 - £330.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Heating; Sizes 20000, 3000 and 5000 mL non-DIN ISO size |
Autoclavable | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11758001
|
DWK Life Sciences
217111708 |
50 mL |
£99.85
Pack of 10 |
Estimated Shipment: 19-04-2024
Log in to see stock. |
|||||
11728021
|
DWK Life Sciences
217112404 |
100 mL |
£77.20
Pack of 10 |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||||
11750744
|
DWK Life Sciences
217113606 |
250 mL |
£90.20
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
11760744
|
DWK Life Sciences
217114405 |
500 mL |
£123.00
Pack of 10 |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||||
11770744
|
DWK Life Sciences
217115401 |
1000 mL |
£181.00
Pack of 10 |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||||
11780744
|
DWK Life Sciences
217116406 |
2000 mL |
£330.00
Pack of 10 |
Estimated Shipment: 19-04-2024
Log in to see stock. |
|||||
10129920
|
DWK Life Sciences
217116809 |
3000 mL |
£63.75
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10363791
|
DWK Life Sciences
217117102 |
4000 mL |
£90.85
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10139920
|
DWK Life Sciences
217117308 |
5000 mL |
£119.00
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
10373791
|
DWK Life Sciences
217117608 |
6000 mL |
£146.00
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
10603341
|
DWK Life Sciences
217118604 |
10,000 mL |
£220.00
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
Description
Abdominal vessels are called flasks in the laboratory and, in addition to beakers, are other universally applicable vessels. They are used e.g. for distillation or for heating liquids. Flasks are available in various designs: narrow and wide neck, with or without ground joint, as round or flat bottom flask. Depending on the application, the required flask with its specific components is selected.
- Flat base means flasks can be set down without a supporting ring
- Fired-on, highly durable white ceramic
Specifications
Borosilicate Glass (3.3) | |
Heating; Sizes 20000, 3000 and 5000 mL non-DIN ISO size | |
Yes |