missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Filtering Flask for DURAN™ Filtering Apparatus, Flask Only

Product Code. 15701738 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
2000 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15701738 2000 mL Each
11699958 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15701738 Supplier DWK Life Sciences Supplier No. 242026306
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The filter unit can be used with an appropriate filter for particle analysis or clear and sterile filtration. Filtration support by vacuum is possible.

One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.

  • Virtually universal applications, as the medium only comes into contact with glass and PTFE
  • The scaled funnel simplifies dosing and analysis (graduated filter head)
  • In addition to the slotted filter plate, porous glass plates with various porosities can also be used thanks to the PTFE plate holder
  • Easy and quick cleaning from both sides thanks to exchangeable filter plates and PTFE adapters
  • The tried-and-tested DURAN™ filtering flask and PTFE hose connection (suitable for tubes with ID 8 mm) enable safe working in the laboratory
  • The replaceable plates and the PTFE adapter in conjunction with the clamp enable rapid changing of porosities or replacement of filters
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY

Specifications

Description Filtering flask with standard ground joint 45/40, 2000 mL
Capacity (Metric) 2000 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Coarse and fine filtration; Filtration without filter paper; Filtering; Residue analysis; Analytical filtration; Purification of aqueous and organic solutions; Particulate contamination analysis; Testing for bacterial contamination; Suitable for GL 14 screw cap (Cat. No. 29 227 05 08)
No. per Pack 1/Pk.
Thread Size GL14
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.