Learn More
DWK Life Sciences DURAN™ Filter Apparatus, Witt Type, complete, with interchangeable lid, and KECK™ assembly set, standard ground joint 45/40
Product Code. p-7189213 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric):
100 mm
150 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10414081 150 mm Each
10570012 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10414081 Supplier DWK Life Sciences Supplier No. 247315703
Only null left
Add to basket
Add to basket
The Witt filtration apparatus is used for solid-liquid filtration in which the filtrate is subsequently further processed.
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
  • Tight closure due to standard ground joint
  • Wide rough-ground tubulature
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (3.3), PBT, Polypropylene, VMQ, EPDM
For Use With (Application) Filtering; Preferably collecting the filtrate; Filtration at reduced pressure; Vacuum filtration; Safe working under vacuum
Quantity 1 Each
Standard Taper Joint NS 45/40
Diameter (Metric) 150 mm