Learn More
Agilent BioTek Dual Reagent Injector
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
LFX, Cytation
Unit Size:
Each
Description
- Accessory required
- Includes 2 syringes, 2 reagent bottles, 2 bottle holders, and inlet tubing
Specifications
Specifications
| Product Type | Dual Reagent Injector |
| For Use With (Equipment) | LFX, Cytation |