missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Dual Casting Base for omniPAGE Maxi Gel System

Product Code. 11854672 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
omniPAGE Maxi Gel System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11854672 omniPAGE Maxi Gel System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11854672
  • ONLINE EXCLUSIVE PRICE
    £158.00
    £150.55 / Pack of 1
    Save £7.45
    4% Off
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Used for high resolution protein and nucleic acid separations.

  • Rapid gel casting and loading

Specifications

For Use With (Equipment) omniPAGE Maxi Gel System
Type Dual Casting Base
Length (Metric) Gel 20 cm
Width (English) Gel 7.87 in.
Width (Metric) Gel 20 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.