missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford DTT
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
100 g
Unit Size:
100g
10g
1g
25g
50g
5g
Description
Protective reagent for maintaining -SH groups in reduced state.
Protective reagent for maintaining -SH groups in reduced state. DTT permeates the cell membrane protecting protein sulfhydryls thereby restoring enzyme activity lost by oxidation of the groups.
Specifications
Specifications
| CAS | 12-3-3483 |
| Molecular Formula | C4H10O2S2 |
| Quantity | 50 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction