missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DTL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111035
This item is not returnable.
View return policy
Description
Immunogen sequence: AGDLPLPSNT PTFSIKTSPA KARSPINRRG SVSSVSPKPP SSFKMSIRNW VTRTPSSSPP ITPPASETKI MSPRKALIPV SQKSSQA Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 86%.
Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control, DNA damage response and translesion DNA synthesis. The DCX(DTL) complex, also named CRL4(CDT2) complex, mediates the polyubiquitination and subsequent degradation of CDT1 and CDKN1A/p21(CIP1). CDT1 degradation in response to DNA damage is necessary to ensure proper cell cycle regulation of DNA replication. CDKN1A/p21(CIP1) degradation during S phase or following UV irradiation is essential to control replication licensing. Most substrates require their interaction with PCNA for their polyubiquitination: substrates interact with PCNA via their PIP-box, and those containing the 'K+4' motif in the PIP box, recruit the DCX(DTL) complex, leading to their degradation. In undamaged proliferating cells, the DCX(DTL) complex also promotes the 'Lys-164' monoubiquitination of PCNA, thereby being involved in PCNA-dependent translesion DNA synthesis.
Specifications
| DTL | |
| Polyclonal | |
| Unconjugated | |
| DTL | |
| 2810047L02Rik; 5730564G15Rik; CDT2; CDW1; DCAF2; DDB1 and CUL4 associated factor 2; DDB1- and CUL4-associated factor 2; denticleless E3 ubiquitin protein ligase homolog; denticleless E3 ubiquitin protein ligase homolog (Drosophila); denticleless homolog; denticleless homolog (Drosophila); denticleless protein homolog; DTL; L2dtl; lethal(2) denticleless protein homolog; Meth A RAMP; Meth A retinoic acid-regulated nuclear matrix-associated protein; RA regulated nuclear matrix associated protein; RAMP; RA-regulated nuclear matrix-associated protein; Retinoic acid-regulated nuclear matrix-associated protein; RGD1310439 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51514 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZJ0 | |
| DTL | |
| Recombinant Protein corresonding to Human DTL. Recombinant protein control fragment (Product #RP-105875). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction