missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dry-Ready™ RT-LAMP Kit

Product Code. 31722824
Change view
Click to view available options
Quantity:
200 Reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31722824 200 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31722824 Supplier Invitrogen™ Supplier No. A40006907
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Experience the simplicity and efficiency of the air drying process by gently heating materials at moderate temperatures in a simple ventilated oven.

The Dry-Ready RT-LAMP Kit is a complete solution for reverse transcription loop-mediated isothermal amplification (RT-LAMP)-based assay air drying. The kit includes all required components: Dry-Ready SuperScript IV Reverse Transcriptase, Dry-Ready Bst DNA Polymerase, Dry-Ready dNTP Mix, and 10X Dry-Ready RT-LAMP Reaction Buffer to facilitate pre- and post-drying procedures.

Features of the Dry-Ready RT-LAMP Kit include:

  • All required components for RT-LAMP assay drying and downstream applications
  • Rapid RNA conversion in RT-LAMP reaction using SuperScript IV Reverse Transcriptase
  • Robust Bst DNA Polymerase with enhanced speed and sensitivity, engineered for fast and efficient amplification in RT-LAMP

The Dry-Ready RT-LAMP Kit includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance.

The Dry-Ready kit is optimized for the air drying process. Notably, air drying is a simple process that does not impair functionality of the assay. On the contrary, it help provides flexibility on the initial volume of sample and increases sensitivity, resulting in a exceptional stabilization process for point-of-care applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type DNA Amplification Kit
Form Liquid
For Use With (Application) RT-LAMP, LAMP
Polymerase Bst DNA Polymerase
Quantity 200 Reactions
Research Category Nucleic Acid Amplification
Content And Storage

•Dry-Ready™ Bst DNA Polymerase
•Dry-Ready™ SuperScript™ IV Reverse Transcriptase
•Dry-Ready™ dNTP Mix
•5X Dry-Ready™ Reaction Buffer
•MgCl2 Solution
•Store at -15 to -25°C

No. of Reactions 200 Reactions
Optimal Reaction Temperature 65°C
Reverse Transcriptase SuperScript™ IV Reverse Transcriptase
Reaction Time ≤10 Min.
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.