missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Dropping Funnels

Manufactured of Borosilicate Glass. DURAN™ Dropping Funnels include stopcocks.

Brand:  Duran™ 241243707

Code : A0

Additional Details : Weight : 0.22000kg

 View more versions of this product

Product Code. 10059990

  • £136.00 / Each

Estimated Shipment
14-01-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

Yes
Dropping funnel
320 mm
5mL
250 mL
Videos
Documents