missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - De Tube Hydrochloric Acid

For making spot measurments for various gases and vapors

Brand:  Dräger™ 8101681

Code : NEW

Additional Details : Weight : 0.08820kg


 View more versions of this product

Product Code. 10252581

  • £108.00 / Pack of 10
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
Specifications

Specifications

Hydrochloric Acid, Nitric Acid 1/a
1 to 10ppm
10
41°F to 104°F
15mg/L
10/20
1.5 to 3 min.
5° to 40°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrochloric Acid / Nitric Acid 1/a

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.

Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid, nitric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers