missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPYSL3 Recombinant Rabbit Monoclonal Antibody (24GB3720)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | DPYSL3 |
|---|---|
| Clone | 24GB3720 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275462
![]() |
Invitrogen™
MA557015 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276615
|
Invitrogen™
MA557016 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DPYSL3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.Specifications
| DPYSL3 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| DPYSL3 | |
| Collapsin response mediator protein 4; collapsin response mediator protein 4 long; Crmp4; CRMP-4; dihydropyrimidinase like 3; dihydropyrimidinase-like 3; dihydropyrimidinase-related protein 3; Dpysl3; DRP3; DRP-3; LCRMP; testicular secretory protein Li 7; TOAD-64/Ulip/CRMP; TOAD-64/Ulip/CRMP protein 4b; TUC4; tuc-4; TUC-4b; ULIP; Ulip1; ULIP-1; Unc-33-like phosphoprotein 1 | |
| DPYSL3 | |
| Primary | |
| Affinity chromatography |
| 24GB3720 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14195, Q62188, Q62952 | |
| 1809, 22240, 25418 | |
| A synthesized peptide derived from human CRMP4 (aa 450-500). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title