missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPYSL3 Recombinant Rabbit Monoclonal Antibody (24GB3720)

Description
DPYSL3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.
Specifications
Specifications
| Antigen | DPYSL3 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 24GB3720 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | DPYSL3 |
| Gene Accession No. | Q14195, Q62188, Q62952 |
| Gene Alias | Collapsin response mediator protein 4; collapsin response mediator protein 4 long; Crmp4; CRMP-4; dihydropyrimidinase like 3; dihydropyrimidinase-like 3; dihydropyrimidinase-related protein 3; Dpysl3; DRP3; DRP-3; LCRMP; testicular secretory protein Li 7; TOAD-64/Ulip/CRMP; TOAD-64/Ulip/CRMP protein 4b; TUC4; tuc-4; TUC-4b; ULIP; Ulip1; ULIP-1; Unc-33-like phosphoprotein 1 |
| Gene Symbols | DPYSL3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?