missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPP3 Recombinant Rabbit Monoclonal Antibody (23GB5190), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | DPP3 |
|---|---|
| Clone | 23GB5190 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088894
![]() |
Invitrogen™
MA553860 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089240
|
Invitrogen™
MA553861 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DPP3 Recombinant Monoclonal antibody specifically detects DPP3 in Human, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| DPP3 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| O55096, Q9NY33 | |
| 10072, 114591 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5190 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4930533O14Rik; C86324; Dipeptidyl aminopeptidase III; dipeptidyl arylamidase III; Dipeptidyl peptidase 3; dipeptidyl peptidase III; dipeptidylpeptidase 3; dipeptidyl-peptidase 3; dipeptidylpeptidase III; dipeptidyl-peptidase III; DPP III; Dpp3; DPPIII; Enkephalinase B; hCG_2016942 | |
| A synthesized peptide derived from human DPP3 (700-737AA). | |
| Primary | |
| -20°C | |
| DPP3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title