missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPP3 Recombinant Rabbit Monoclonal Antibody (23GB5190), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA553860

 View more versions of this product

Product Code. 30088894

  • £485.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DPP3
Recombinant Monoclonal
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O55096, Q9NY33
DPP3
A synthesized peptide derived from human DPP3 (700-737AA).
100 μL
Primary
Human, Rat
Antibody
IgG
Flow Cytometry, Western Blot
23GB5190
Unconjugated
DPP3
4930533O14Rik; C86324; Dipeptidyl aminopeptidase III; dipeptidyl arylamidase III; Dipeptidyl peptidase 3; dipeptidyl peptidase III; dipeptidylpeptidase 3; dipeptidyl-peptidase 3; dipeptidylpeptidase III; dipeptidyl-peptidase III; DPP III; Dpp3; DPPIII; Enkephalinase B; hCG_2016942
Rabbit
Affinity chromatography
RUO
10072, 114591
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.