missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | DP1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DP1 Polyclonal specifically detects DP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| DP1 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS buffer, 2% sucrose | |
| 7027 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DP1E2F dimerization partner 1, DRTF1Dp-1, DRTF1-polypeptide 1, E2F-related transcription factor, transcription factor Dp-1 | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human DP1 (NP_009042). Peptide sequence VIGTPQRPAASNTLVVGSPHTPSTHFASQNQPSDSSPWSAGKRNRKGEKN | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title