missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline monohydrate, Thermo Scientific™

Doxycycline monohydrate, CAS # 17086-28-1, is the hydrated form of the tetracycline class antibiotic doxycycline.

£59.81 - £227.99

Chemical Identifiers

CAS 17086-28-1
Molecular Formula C22H26N2O9
Molecular Weight (g/mol) 462.455
MDL Number MFCD02682958
InChI Key FZKWRPSUNUOXKJ-CVHRZJFOSA-N
PubChem CID 54684461
IUPAC Name (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
SMILES CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15435979
SDS View Documents
Thermo Scientific Alfa Aesar
J63805.06
5g
ONLINE EXCLUSIVE PRICE
£61.10
£59.81 / 5g
Save £1.29
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15445979
SDS View Documents
Thermo Scientific Alfa Aesar
J63805.14
25g
ONLINE EXCLUSIVE PRICE
£233.00
£227.99 / 25g
Save £5.01
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Doxycycline monohydrate is a semi-synthetic tetracycline antibiotic with broad-spectrum activity
• It is a tetracycline with the 5beta-hydrogen group replaced by a hydroxy group
• Doxycycline monohydrate acts by binding the 30S ribosomal subunit and preventing polypeptide formation
• Doxycycline monohydrate may also act on the 50S ribosomal subunit, is a known inhibitor of collagenases, and has anti-inflammatory properties
Application
• Doxycycline can be used for study of cancers, infections, and antibiotic resistance in laboratory settings
• Doxycycline can be used experimentally to control TET genes and induce expression of genes of interest in vivo and in vitro
Specifications

Chemical Identifiers

17086-28-1
462.455
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
C22H26N2O9
MFCD02682958
54684461
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O

Specifications

17086-28-1
C22H26N2O9
14,3440
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
54684461
Doxycycline monohydrate
Yellow
MFCD02682958
Soluble in ethanol or methanol; Sparingly soluble in water;Soluble in dilute acids,alkali hydroxides and dimethyl sulfoxide.
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O
462.455
462.46 (444.44 Anhydrous)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P201-P202-P281-P308+P313-P501c

H361d

missing translation for 'tsca' : No

Recommended Storage : Keep cold

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Consejos de prudencia
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documentos

Documentos

Product Certifications

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado