missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline monohydrate, Thermo Scientific™

Doxycycline monohydrate, CAS # 17086-28-1, is the hydrated form of the tetracycline class antibiotic doxycycline.

Brand:  Thermo Scientific Alfa Aesar J63805.14


Quantity 25g

 View more versions of this product

Product Code. 15445979

  • ONLINE EXCLUSIVE PRICE
    £233.00
    £227.99 / 25g
    Save £5.01
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Doxycycline monohydrate is a semi-synthetic tetracycline antibiotic with broad-spectrum activity
• It is a tetracycline with the 5beta-hydrogen group replaced by a hydroxy group
• Doxycycline monohydrate acts by binding the 30S ribosomal subunit and preventing polypeptide formation
• Doxycycline monohydrate may also act on the 50S ribosomal subunit, is a known inhibitor of collagenases, and has anti-inflammatory properties
Application
• Doxycycline can be used for study of cancers, infections, and antibiotic resistance in laboratory settings
• Doxycycline can be used experimentally to control TET genes and induce expression of genes of interest in vivo and in vitro
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

17086-28-1
462.455
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
C22H26N2O9
MFCD02682958
54684461
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O

Specifications

17086-28-1
25g
MFCD02682958
Soluble in ethanol or methanol; Sparingly soluble in water;Soluble in dilute acids,alkali hydroxides and dimethyl sulfoxide.
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O
462.455
462.46 (444.44 Anhydrous)
Yellow
C22H26N2O9
14,3440
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
54684461
Doxycycline monohydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.