missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Double Tray Rack for Upright Freezers, Standard

Trays designed for applications where the samples are either not stored in boxes or has odd sizes.

Brand:  Tenak™ TE29320

Additional Details : Weight : 0.55000kg

 View more versions of this product

Product Code. 12487000

  • £142.00 / Each
Estimated Shipment: 24-07-2024
to see stock.
Add to basket

Description

Description

  • Double trays are a rack with two drawers including a lid with a handle on the front
  • Double trays will fit into any freezer
  • Constructed of stainless steel
Specifications

Specifications

Handle and Lid
Upright Freezers
139 mm
Stainless Steel
560 x 139 x 170 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers