missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Magnetic Stir Bar, Double-sided crosshead

Product Code. p-8007713 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
10 mm
14 mm
17 mm
22 mm
30 mm
35 mm
40 mm
60 mm
Unit Size:
Each
Pack of 10
Pack of 5

Product Code. 11838892

Only null left
Add to basket
Edge
Add to basket

Designed for use with tube-like containers yet very effective as general stirrers. The PTFE-coated magnet stirrers include a crosshead on both sides of the stirrer for greater turbulence.

  • Stable, general-purpose magnetic stirrer
  • Designed for use with tube-like containers yet very effective as general stirrers
  • Double-sided crossheads provider greater turbulence during mixing
  • Autoclavable

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Height (Metric) 8mm
Coating PTFE
Diameter (Metric) 10 mm
Quantity 5/Pk.
Type PTFE Stirrer Bars Crosshead
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.