missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Dosing/Measuring Spoon Set
BIOPHARMA

Product Code. 16525260 Shop All Bürkle Products
Click to view available options
Color:
Blue
White
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 16525260

Brand: Buerkle™ 53784120

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Spoon set can be used to precisely measure and dose small amounts of powder, granules, and liquids.

  • For precise measuring, excess medium is stripped off the upper edge of the spoon
  • The content volume is engraved on the handle
  • The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, allowing for good emptying of small remaining amounts
  • The flat base makes it possible to set the volumetric spoon down without spilling the content
  • An eyelet in the end of the handle can be used to hang up a spoon or to bundle several together
  • Content volume: 0.5, 1.0, 2.5, 5.0, 10, 15, 25, and 50 mL
  • Set of volumetric spoons with eight sizes
  • With stripper edge
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100
Description Dosing spoons with stripper edge for exact measuring
Sterility Non-sterile
Capacity (Metric) 0.5 to 50 mL
Color White
Certifications/Compliance Food Contact, Reach, RoHS
Disposable Single-use
Product Type Spoon
For Use With (Application) Especially for The Food, Animal Feed and Pharmaceutical Industries
Material Polystyrene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.