Learn More
KARTELL DOSILAB
Dosilab is a new series of adjustable volume bottle top dispenser for dosing chemicals with precision, accuracy and inexpensiveness. The six models of the new line are simple to use and easy to disassemble to perform common cleaning and maintenance operations.
Brand: KARTELL 8308500
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Completely autoclavable at +121°C without need for disassembly.
- PTFE piston with silicone O-ring ensuring a high degree of accuracy.
- Internal cylinder in borosilicate glass protected by PP sheath.
- Valve housing: PTFE body in a single piece ensuring high chemical resistance.
- Accuracy and Reproducibility: according to the ISO 8655 standards.
- Compatibility: excellent with all reagents including organic solvents and concentrated acids, except hydrofluoric acid (HF).
- Recalibration: simple and quick with its special accessory.
- All the internal components that during use are in contact with liquid solutions are made of PTFE, FEP and borosilicate glass. They therefore possess excellent compatibility with all reagents (even organic solvents and concentrated acids), except for hydrofluoric acid.
Specifications
± 0.150 mL | |
Autoclavable | |
White | |
FEP, PTFE, Borosilicate Glass, Polypropylene | |
Analog | |
0.5 mL |
Adjustable Volume Dispenser | |
ISO 8655 | |
1 | |
30 mL | |
Reagent Bottle | |
1.01 oz. |