missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL DOSILAB
Dosilab is a new series of adjustable volume bottle top dispenser for dosing chemicals with precision, accuracy and inexpensiveness. The six models of the new line are simple to use and easy to disassemble to perform common cleaning and maintenance operations.
£291.00 - £486.00
Specifications
Product Type | Adjustable Volume Dispenser |
---|---|
Autoclavable | Autoclavable |
Color | White |
Channels | 1 |
Display | Analog |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
12860933
|
KARTELL
8308200 |
2.5 mL |
£291.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12870933
|
KARTELL
8308300 |
5 mL |
£291.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12890933
|
KARTELL
8308400 |
10 mL |
£291.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12880933
|
KARTELL
8308500 |
30 mL |
£371.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12800943
|
KARTELL
8308600 |
60 mL |
£371.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12810943
|
KARTELL
8308700 |
100 mL |
£486.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Description
- Completely autoclavable at +121°C without need for disassembly.
- PTFE piston with silicone O-ring ensuring a high degree of accuracy.
- Internal cylinder in borosilicate glass protected by PP sheath.
- Valve housing: PTFE body in a single piece ensuring high chemical resistance.
- Accuracy and Reproducibility: according to the ISO 8655 standards.
- Compatibility: excellent with all reagents including organic solvents and concentrated acids, except hydrofluoric acid (HF).
- Recalibration: simple and quick with its special accessory.
- All the internal components that during use are in contact with liquid solutions are made of PTFE, FEP and borosilicate glass. They therefore possess excellent compatibility with all reagents (even organic solvents and concentrated acids), except for hydrofluoric acid.
Specifications
Adjustable Volume Dispenser | |
White | |
Analog |
Autoclavable | |
1 |