missing translation for 'onlineSavingsMsg'
Learn More

KARTELL DOSILAB

Dosilab is a new series of adjustable volume bottle top dispenser for dosing chemicals with precision, accuracy and inexpensiveness. The six models of the new line are simple to use and easy to disassemble to perform common cleaning and maintenance operations.

Brand:  KARTELL 8308300

Code : NEW

Additional Details : Weight : 0.74000kg

 View more versions of this product

Product Code. 12870933

  • £291.00 / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • Completely autoclavable at +121°C without need for disassembly.
  • PTFE piston with silicone O-ring ensuring a high degree of accuracy.
  • Internal cylinder in borosilicate glass protected by PP sheath.
  • Valve housing: PTFE body in a single piece ensuring high chemical resistance.
  • Accuracy and Reproducibility: according to the ISO 8655 standards.
  • Compatibility: excellent with all reagents including organic solvents and concentrated acids, except hydrofluoric acid (HF).
  • Recalibration: simple and quick with its special accessory.
  • All the internal components that during use are in contact with liquid solutions are made of PTFE, FEP and borosilicate glass. They therefore possess excellent compatibility with all reagents (even organic solvents and concentrated acids), except for hydrofluoric acid.
Specifications

Specifications

± 0.025 mL
Autoclavable
White
FEP, PTFE, Borosilicate Glass, Polypropylene
Analog
0.1 mL
Adjustable Volume Dispenser
ISO 8655
1
5 mL
Reagent Bottle
0.17 oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers