missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doppel Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | Doppel |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633782
|
Novus Biologicals
NBP2-92485-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639221
|
Novus Biologicals
NBP2-92485-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
Doppel Polyclonal antibody specifically detects Doppel in Human, Mouse, Rat samples. It is validated for Western BlotEspecificaciones
| Doppel | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neurodegeneration, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 23627 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| dJ1068H6.4, DOPPEL, DPLprion gene complex, downstream, MGC41841, Prion protein 2, prion protein 2 (dublet), PrPLPprion-like protein doppel | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-160 of human PRND (NP_036541.2). HLSAVQTRGIKHRIKWNRKALPSTAQITEAQVAENRPGAFIKQGRKLDIDFGAEGNRYYEANYWQFPDGIHYNGCSEANVTKEAFVTGCINATQAANQGEFQKPDNKLHQQVLWRLVQELCSLKHCEFWLERGAGLRVTMH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto