missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doppel Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92485-0.02ml
This item is not returnable.
View return policy
Description
Doppel Polyclonal antibody specifically detects Doppel in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Doppel | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| dJ1068H6.4, DOPPEL, DPLprion gene complex, downstream, MGC41841, Prion protein 2, prion protein 2 (dublet), PrPLPprion-like protein doppel | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-160 of human PRND (NP_036541.2). HLSAVQTRGIKHRIKWNRKALPSTAQITEAQVAENRPGAFIKQGRKLDIDFGAEGNRYYEANYWQFPDGIHYNGCSEANVTKEAFVTGCINATQAANQGEFQKPDNKLHQQVLWRLVQELCSLKHCEFWLERGAGLRVTMH | |
| 0.02 mL | |
| Neurodegeneration, Neuroscience | |
| 23627 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction