missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dopamine Transporter Recombinant Rabbit Monoclonal Antibody (24GB2160)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen Dopamine Transporter
Clone 24GB2160
Concentration 0.40 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276616
GREENER_CHOICE
View Documents
Invitrogen™
MA557261
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30274984
View Documents
Invitrogen™
MA557262
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dopamine Transporter Recombinant Monoclonal Antibody for Western Blot, Flow

The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
Specifications

Specifications

Dopamine Transporter
0.40 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q01959
6531
A synthesized peptide derived from human Dopamine Transporter (aa 150-215).
Antibody
24GB2160
-20°C
Recombinant Monoclonal
Liquid
IgG
Human
Slc6a3
DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3
Slc6a3
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.