missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dopamine Transporter Recombinant Rabbit Monoclonal Antibody (24GB2160)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557261
This item is not returnable.
View return policy
Description
Dopamine Transporter Recombinant Monoclonal Antibody for Western Blot, Flow
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
Specifications
| Dopamine Transporter | |
| Recombinant Monoclonal | |
| 0.40 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q01959 | |
| Slc6a3 | |
| A synthesized peptide derived from human Dopamine Transporter (aa 150-215). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2160 | |
| Unconjugated | |
| Slc6a3 | |
| DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6531 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction