missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOC2B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00 - £397.00
Specifications
| Antigen | DOC2B |
|---|---|
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18238671
|
Novus Biologicals
NBP1-03473-0.1MG |
0.1 mg |
£134.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18203650
|
Novus Biologicals
NBP1-03473 |
0.5 mg |
£397.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DOC2B Polyclonal specifically detects DOC2B in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DOC2B | |
| Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat, Mouse | |
| DOC2BL, double C2-like domains, beta | |
| DOC2B | |
| Primary |
| LYOPH | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| P70610 | |
| 8447 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title