missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOC2B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03473
This item is not returnable.
View return policy
Description
DOC2B Polyclonal specifically detects DOC2B in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DOC2B | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from PBS with No Preservative | |
| DOC2BL, double C2-like domains, beta | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml | |
| P70610 | |
| DOC2B | |
| A synthetic peptide from internal region of rat DOC2B conjugated to an immunogenic carrier protein was used as the antigen. | |
| 0.5 mg | |
| Neuroscience | |
| 8447 | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction