missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOC2B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03473-0.1MG
This item is not returnable.
View return policy
Description
DOC2B Polyclonal specifically detects DOC2B in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DOC2B | |
| Polyclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| DOC2BL, double C2-like domains, beta | |
| Rabbit | |
| IgG purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml | |
| P70610 | |
| DOC2B | |
| A synthetic peptide from the n-terminal of rat Doc2b ( Doc2-beta, Double C2-like domain-containing protein beta) conjugated to an immunogenic carrier protein was used as the immunogen. | |
| 0.1 mg | |
| Neuroscience | |
| 8447 | |
| Rat, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction