missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOC1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£177.00 - £366.00
Specifications
| Antigen | DOC1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647120
|
Novus Biologicals
NBP2-92390-0.02ml |
0.02 mL |
£177.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678200
|
Novus Biologicals
NBP2-92390-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DOC1 Polyclonal antibody specifically detects DOC1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| DOC1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cardiovascular Biology | |
| PBS with 50% glycerol, pH7.3. | |
| 11259 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 90 kDa GPBP-interacting protein, downregulated in ovarian cancer 1 81 kDa isoform, filamin A interacting protein 1-like, GIP130a, GIP130b, GIP130c | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human FILIP1L (NP_878913.2). MRSRGSDTEGSAQKKFPRHTKGHSFQGPKNMKHRQQDKDSPSESDVILPCPKAEKPHSGNGHQAEDLSRDDLLFLLSILEGELQARDEVIGILKAEKMDLALLEAQYGFVTPKKVLEALQRDAFQAKSTPWQEDIYEKPMNELDKVVEKHKESYRRILGQLLVAEKSRRQTILELEEEKRKHKEYMEKSDEFICLLEQEC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title