missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOC1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92390-0.1ml
This item is not returnable.
View return policy
Description
DOC1 Polyclonal antibody specifically detects DOC1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DOC1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 90 kDa GPBP-interacting protein, downregulated in ovarian cancer 1 81 kDa isoform, filamin A interacting protein 1-like, GIP130a, GIP130b, GIP130c | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human FILIP1L (NP_878913.2). MRSRGSDTEGSAQKKFPRHTKGHSFQGPKNMKHRQQDKDSPSESDVILPCPKAEKPHSGNGHQAEDLSRDDLLFLLSILEGELQARDEVIGILKAEKMDLALLEAQYGFVTPKKVLEALQRDAFQAKSTPWQEDIYEKPMNELDKVVEKHKESYRRILGQLLVAEKSRRQTILELEEEKRKHKEYMEKSDEFICLLEQEC | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology | |
| 11259 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction