missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNTTIP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | DNTTIP1 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DNTTIP1 Polyclonal antibody specifically detects DNTTIP1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DNTTIP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS with 50% glycerol, pH7.3. | |
| 116092 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C20orf167, chromosome 20 open reading frame 167, deoxynucleotidyltransferase terminal-interacting protein 1, deoxynucleotidyltransferase, terminal, interacting protein 1, dJ447F3.4, TDIF1, TdT binding protein, TdT-interacting factor 1, Terminal deoxynucleotidyltransferase-interacting factor 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-329 of human DNTTIP1 (NP_443183.1). MGATGDAEQPRGPSGAERGGLELGDAGAAGQLVLTNPWNIMIKHRQVQRRGRRSQMTTSFTDPAISMDLLRAVLQPSINEEIQTVFNKYMKFFQKAALNVRDNVGEEVDAEQLIQEACRSCLEQAKLLFSDGEKVIPRLTHELPGIKRGRQAEEECAHRGSPLPKKRKGRPPGHILSSDRAAAGMVWKPKSCEPIRREGPKWDPARLNESTTFVLGSRANKALGMGGTRGRIYIKHPHLFKYAADPQDKHWLAEQHHMRATGGKMAYLLIEEDIRDLAASDDYRGCLDLKLEELKSFVLPSWMVEKMRKYMETLRTENEHRAVEAPPQT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title