missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNTTIP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92249-0.02ml
This item is not returnable.
View return policy
Description
DNTTIP1 Polyclonal antibody specifically detects DNTTIP1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DNTTIP1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| C20orf167, chromosome 20 open reading frame 167, deoxynucleotidyltransferase terminal-interacting protein 1, deoxynucleotidyltransferase, terminal, interacting protein 1, dJ447F3.4, TDIF1, TdT binding protein, TdT-interacting factor 1, Terminal deoxynucleotidyltransferase-interacting factor 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-329 of human DNTTIP1 (NP_443183.1). MGATGDAEQPRGPSGAERGGLELGDAGAAGQLVLTNPWNIMIKHRQVQRRGRRSQMTTSFTDPAISMDLLRAVLQPSINEEIQTVFNKYMKFFQKAALNVRDNVGEEVDAEQLIQEACRSCLEQAKLLFSDGEKVIPRLTHELPGIKRGRQAEEECAHRGSPLPKKRKGRPPGHILSSDRAAAGMVWKPKSCEPIRREGPKWDPARLNESTTFVLGSRANKALGMGGTRGRIYIKHPHLFKYAADPQDKHWLAEQHHMRATGGKMAYLLIEEDIRDLAASDDYRGCLDLKLEELKSFVLPSWMVEKMRKYMETLRTENEHRAVEAPPQT | |
| 0.02 mL | |
| DNA replication Transcription Translation and Splicing | |
| 116092 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction