missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNP-X, SE, 6-(2,4-Dinitrophenyl)aminohexanoic Acid, Succinimidyl Ester

Product Code. 11510266
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11510266 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11510266 Supplier Invitrogen™ Supplier No. D2248
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

DNP-X, SE, 6-(2,4-Dinitrophenyl)aminohexanoic Acid, Succinimidyl Ester

The amine-reactive DNP-X succinimidyl ester can be used bioconjugates that can be detected with anti-dinitrophenyl (anti-DNP) antibodies. This reactive hapten contains an additional seven-atom aminohexanoyl spacer ('X') between the fluorophore and the succinimidyl ester group. This spacer helps to separate increase accessibility to anti-DNP antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type DNP-X Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye DNP
Label Type Biotin and Other Haptens

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.