missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3B Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £318.00
Specifications
| Antigen | DNMT3B |
|---|---|
| Dilution | Western Blot 1:1000, ELISA 1:100-1:500 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18307985
|
Novus Biologicals
NBP3-18646-25UL |
25 μg |
£139.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18306255
|
Novus Biologicals
NBP3-18646-100UL |
100 μg |
£318.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNMT3B Polyclonal antibody specifically detects DNMT3B in Human, Mouse samples. It is validated for Western Blot, ELISASpecifications
| DNMT3B | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Chromatin Research, DNA Repair, Epigenetics | |
| Whole antiserum | |
| DNA (cytosine-5-)-methyltransferase 3 beta, DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, Dnmt3b, EC 2.1.1.37, ICF, M.HsaIIIB | |
| Polyclonal antibody raised in rabbit against mouse DNMT3B (DNA methyltransferase 3B), using a KLH-conjugated synthetic peptide containing a sequence from the N-terminal part of the protein (1). | |
| Whole Antiserum |
| Western Blot 1:1000, ELISA 1:100-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse | |
| Q9UBC3 | |
| 1789 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title