missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3B Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18646-100UL
This item is not returnable.
View return policy
Description
DNMT3B Polyclonal antibody specifically detects DNMT3B in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| DNMT3B | |
| Polyclonal | |
| Western Blot 1:1000, ELISA 1:100-1:500 | |
| Q9UBC3 | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antisera |
| Western Blot, ELISA | |
| Unconjugated | |
| Whole antiserum | |
| DNA (cytosine-5-)-methyltransferase 3 beta, DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, Dnmt3b, EC 2.1.1.37, ICF, M.HsaIIIB | |
| Polyclonal antibody raised in rabbit against mouse DNMT3B (DNA methyltransferase 3B), using a KLH-conjugated synthetic peptide containing a sequence from the N-terminal part of the protein (1). | |
| 100 μg | |
| Cancer, Chromatin Research, DNA Repair, Epigenetics | |
| 1789 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction