missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3A Antibody (64B1446) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£229.00 - £451.00
Specifications
| Antigen | DNMT3A |
|---|---|
| Clone | 64B1446 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot, Chromatin Immunoprecipitation reported in scientific literature (PMID 24623306), Flow Cytometry 1 ug per million cells, Immunohistochemistry reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml. Use reported in scientific literature (PMID 22134929), Immunohistochemistry-Frozen reported in scientific literature (PMID 22134929), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 26507142), Chromatin Immunoprecipitation (ChIP), CyTOF-ready |
| Applications | Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18748133
|
Novus Biologicals
NB120-13888SS |
0.025 mg |
£229.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18799003
|
Novus Biologicals
NB120-13888 |
0.1 mg |
£451.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNMT3A Monoclonal specifically detects DNMT3A in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Whole-Mount, CyTOF.Specifications
| DNMT3A | |
| 1.0 mg/mL | |
| Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
| Q9Y6K1 | |
| 1788 | |
| This DNMT3A antibody was raised against bacteria expressed recombinant mouse Dnmt3a. The epitope was found to lie near the C-terminus (a.a. 705-908), see Chen et (2002) for details. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 64B1446 | |
| Western Blot, Chromatin Immunoprecipitation reported in scientific literature (PMID 24623306), Flow Cytometry 1 ug per million cells, Immunohistochemistry reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml. Use reported in scientific literature (PMID 22134929), Immunohistochemistry-Frozen reported in scientific literature (PMID 22134929), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 26507142), Chromatin Immunoprecipitation (ChIP), CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 64B1446 clone, DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA | |
| DNMT3A | |
| IgG1 κ | |
| Protein G purified | |
| Cross-reacts with Human (weakly) and Mouse. Not yet tested in other species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title