missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3A Antibody (64B1446) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-13888SS
This item is not returnable.
View return policy
Description
DNMT3A Monoclonal specifically detects DNMT3A in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Whole-Mount, CyTOF.
Specifications
| DNMT3A | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, Chromatin Immunoprecipitation reported in scientific literature (PMID 24623306), Flow Cytometry 1 ug per million cells, Immunohistochemistry reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml. Use reported in scientific literature (PMID 22134929), Immunohistochemistry-Frozen reported in scientific literature (PMID 22134929), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 26507142), Chromatin Immunoprecipitation (ChIP), CyTOF-ready | |
| 64B1446 clone, DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cross-reacts with Human (weakly) and Mouse. Not yet tested in other species. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 64B1446 | |
| Unconjugated | |
| Q9Y6K1 | |
| DNMT3A | |
| This DNMT3A antibody was raised against bacteria expressed recombinant mouse Dnmt3a. The epitope was found to lie near the C-terminus (a.a. 705-908), see Chen et (2002) for details. | |
| 0.025 mg | |
| Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
| 1788 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction