missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNALI1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56390
This item is not returnable.
View return policy
Description
DNALI1 Polyclonal specifically detects DNALI1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DNALI1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DNALI1 | |
| Synthetic peptides corresponding to DNALI1(dynein, axonemal, light intermediate chain 1) The peptide sequence was selected from the N terminal of DNALI1. Peptide sequence MVTANKAHTGQGSCWVATLASAMIPPADSLLKYDTPVLVSRNTEKRSPKA. | |
| 100 μL | |
| Signal Transduction | |
| 7802 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| dJ423B22.5, dynein, axonemal, light intermediate chain 1, dynein, axonemal, light intermediate polypeptide 1, hp28axonemal dynein light intermediate polypeptide 1, Inner dynein arm light chain, axonemal, inner dynein arm, homolog of clamydomonas, P28dJ423B22.5 (axonemal dynein light chain (hp28)) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Mouse: 100%; Rabbit: 100%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction