missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48704
This item is not returnable.
View return policy
Description
DNAJC3 Polyclonal antibody specifically detects DNAJC3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| DNAJC3 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| DnaJ (Hsp40) homolog, subfamily C, member 3, HP58, Interferon-induced, double-stranded RNA-activated protein kinase inhibitor, P58FLJ21288, P58IPKProtein kinase inhibitor p58, PRKRIdnaJ homolog subfamily C member 3, Protein kinase inhibitor of 58 kDa, protein-kinase, interferon-inducible double stranded RNA dependent inhibitor | |
| This antibody was developed against a recombinant protein corresponding to amino acids: YKISTLYYQLGDHELSLSEVRECLKLDQDHKRCFAHYKQVKKLNKLIESAEELIRDGRYTDATSKYESVMKTEPSIAEYTVR | |
| 0.1 mL | |
| Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism, Unfolded Protein Response | |
| 5611 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction