Learn More
Invitrogen™ DNAJB6 Monoclonal Antibody (K24018_5D5), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | DNAJB6 |
|---|---|
| Clone | K24018_5D5 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30211253
|
Invitrogen™
MA555688 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30195457
|
Invitrogen™
MA556068 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: QRHASPEDIK KAYRKLALKW HPDKNPENKE EAERKFKQVA EAYEVLSDAK KRDIYDKYGK EGLNGGGGGG SHFDSPFEFG FTFRNPDDVF REFFGGRDPF SFDFFEDPFE DFFGNRRGPR GSRSRGTGSF FSAFSGFPSF GSGFSSFDTG FTSFGSLGHG GLTSFSSTSF GGSGMGNFKS ISTSTKMVNG RKITTKRIVE NGQERVEVEE DGQLKSLTIN GVADDDALAE
DNAJB6 Monoclonal antibody specifically detects DNAJB6 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| DNAJB6 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| DJ4; DnaJ; DnaJ (Hsp40) homolog, subfamily B, member 6; DnaJ heat shock protein family (Hsp40) member B6; dnaJ homolog subfamily B member 6; DNAJB6; dnajb6 {ECO:0000312; DnaJ-like 2 protein; EMBL:AAH78908.1, ECO:0000312; Heat shock protein J2; HHDJ1; Hsj2; HSJ-2; Hsp40 homolog; LGMD1D; LGMD1E; mammalian relative of DnaJ; mDj4; MRJ; MSJ1; MSJ-1; RGD:1308207} | |
| Recombinant human DNAJB6 (immunogen range is 11-240). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| DNAJB6 |
| K24018_5D5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| O54946, O75190 | |
| 10049, 23950 | |
| IgG2a | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.