missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB4 Mouse anti-Human, Unconjugated, Clone: 1F3H7, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DNAJB4 |
|---|---|
| Clone | 1F3H7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17581476
|
Proteintech
67966-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17571476
|
Proteintech
67966-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DNAJB4 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human DNAJB4. This antibody recognizes human, pig, rabbit, mouse antigen. The DNAJB4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| DNAJB4 | |
| Western Blot | |
| Unconjugated | |
| Human, Pig, Rabbit, Mouse | |
| IgG1 |
| 1F3H7 | |
| Monoclonal | |
| Mouse | |
| 11080 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title