Learn More
DNAJB4 Mouse anti-Human, Unconjugated, Clone: 1F3H7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67966-1-IG-150UL
This item is not returnable.
View return policy
Description
The DNAJB4 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human DNAJB4. This antibody recognizes human, pig, rabbit, mouse antigen. The DNAJB4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| DNAJB4 | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 11080 | |
| IgG1 |
| Western Blot | |
| 1F3H7 | |
| Mouse | |
| Primary | |
| Human, Pig, Rabbit, Mouse |