missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJA4 Recombinant Rabbit Monoclonal Antibody (23GB5825), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | DNAJA4 |
|---|---|
| Clone | 23GB5825 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087546
![]() |
Invitrogen™
MA552922 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089221
|
Invitrogen™
MA552923 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAJA4 Recombinant Monoclonal antibody specifically detects DNAJA4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| DNAJA4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8WW22, Q9JMC3 | |
| 300721, 55466, 58233 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5825 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110021L12Rik; AV358213; Dj4; DnaJ (Hsp40) homolog, subfamily A, member 4; DnaJ heat shock protein family (Hsp40) member A4; dnaJ homolog subfamily A member 4; DNAJA4; heat shock protein, DNAJ-like 4; Hsj4; mmDjA4; MST104; MSTP104; PRO1472 | |
| A synthesized peptide derived from human DNAJA4 (360-397AA). | |
| Primary | |
| -20°C | |
| DNAJA4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title