missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJA4 Recombinant Rabbit Monoclonal Antibody (23GB5825), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552922
This item is not returnable.
View return policy
Description
DNAJA4 (DnaJ Heat Shock Protein Family (Hsp40) Member A4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include unfolded protein binding and heat shock protein binding.
Specifications
| DNAJA4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DNAJA4 | |
| 1110021L12Rik; AV358213; Dj4; DnaJ (Hsp40) homolog, subfamily A, member 4; DnaJ heat shock protein family (Hsp40) member A4; dnaJ homolog subfamily A member 4; DNAJA4; heat shock protein, DNAJ-like 4; Hsj4; mmDjA4; MST104; MSTP104; PRO1472 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 300721, 55466, 58233 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5825 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WW22, Q9JMC3 | |
| DNAJA4 | |
| A synthesized peptide derived from human DNAJA4 (360-397AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction