missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Primase small subunit Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£140.00 - £283.00
Specifications
| Antigen | DNA Primase small subunit |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18347876
|
Novus Biologicals
NBP3-15505-20UL |
20 μg |
£140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18344534
|
Novus Biologicals
NBP3-15505-100UL |
100 μg |
£283.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA Primase small subunit Polyclonal antibody specifically detects DNA Primase small subunit in Human, Mouse samples. It is validated for Western BlotSpecifications
| DNA Primase small subunit | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DNA primase 1, DNA primase 49 kDa subunit, DNA primase small subunit, DNA primase subunit 48, EC 2.7.7, EC 2.7.7.-, MGC12308, p49, primase p49 subunit, primase polypeptide 1, 49kDa, primase, DNA, polypeptide 1 (49kDa) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 121-420 of human DNA Primase small subunit (NP_000937.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
| PBS, 50% glycerol, pH7.3 | |
| 5557 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title