missing translation for 'onlineSavingsMsg'
Learn More

DNA Primase small subunit Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18344534 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344534 100 μg 100µL
18347876 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18344534 Supplier Novus Biologicals Supplier No. NBP315505100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNA Primase small subunit Polyclonal antibody specifically detects DNA Primase small subunit in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Primase small subunit
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias DNA primase 1, DNA primase 49 kDa subunit, DNA primase small subunit, DNA primase subunit 48, EC 2.7.7, EC 2.7.7.-, MGC12308, p49, primase p49 subunit, primase polypeptide 1, 49kDa, primase, DNA, polypeptide 1 (49kDa)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 121-420 of human DNA Primase small subunit (NP_000937.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5557
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.